• USA Home
  • 1.25259 - Chromolith® Prep column coupler

EMAIL THIS PAGE TO A FRIEND
1.25259 Supelco

Chromolith® Prep column coupler

Purchase

Properties

Related Categories Analytical/Chromatography, LPLC/TLC/Paper Chromatography, TLC Accessories, Thin Layer Chromatography (TLC)
Quality Level   100
application(s)   HPLC: suitable
storage conditions   no restrictions.

Description

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?