EMAIL THIS PAGE TO A FRIEND
1.44159 Millipore

Isokinetic Probe

for use with APC ErgoTouch Pro/APC ErgoTouch

Purchase

Properties

Related Categories Analytical/Chromatography, LPLC/TLC/Paper Chromatography, TLC Accessories, Thin Layer Chromatography (TLC)
compatibility   for use with APC ErgoTouch Pro/APC ErgoTouch
storage conditions   Store at room temperature.

Description

General description

942602 (APC ErgoTouch Pro/APC ErgoTouch)

Stainless steel

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Active Particle Counting for the Food & Beverage Industry: the APC ErgoTouch Hand-Held Particle Counter

The hygiene standard in a food and beverage production environment is directly linked to the microbiological safety of the finished products. Make sure that your active microbial air sampling solutio...
Keywords: Air sampling, Industries, Sample preparations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?