• USA Home
  • 144170 - Nozzle Set Compressed Gas Adapter, 1 CFM

EMAIL THIS PAGE TO A FRIEND
144170 Millipore

Nozzle Set Compressed Gas Adapter, 1 CFM

for use with APC SMT/APC portable

Purchase

Properties

Related Categories Analytical/Chromatography, LPLC/TLC/Paper Chromatography, Media by Application, Media for Cosmetic Industry, Microbiological Culture Media,
compatibility   for use with APC SMT/APC portable
shipped in   ambient
storage conditions   Store at room temperature.

Description

General description

In conjunction with the APC Compressed Gas Adapter and an APC Airborne Particle Counter this item allows for sampling of compressed gases.

for APC SMT/APC Portable

Packaging

1 SET in Fibre carton

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Active Particle Counting for the Food & Beverage Industry: the APC ErgoTouch Hand-Held Particle Counter

The hygiene standard in a food and beverage production environment is directly linked to the microbiological safety of the finished products. Make sure that your active microbial air sampling solutio...
Keywords: Air sampling, Industries, Sample preparations

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?