• USA Home
  • 168320 - Ammonium Chloride, Molecular Biology Grade - CAS 12125-02-9 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
168320 Millipore

Ammonium Chloride, Molecular Biology Grade - CAS 12125-02-9 - Calbiochem

Synonym: Ammonium Chloride, Molecular Biology Grade - CAS 12125-02-9 - Calbiochem, NH₄Cl

  • CAS Number 12125-02-9

  • Empirical Formula (Hill Notation) H4ClN

  • Molecular Weight 53.49

Purchase

Properties

Quality Level   100
assay   ≥99% (silver nitrate titration)
form   crystalline solid
mfr. no.   Calbiochem®
storage condition   OK to freeze
storage conditions   +15C to +30C
color   off-white
solubility   water: 1 M
foreign activity   DNases, proteases, RNases, none detected
shipped in   ambient
InChI   1S/ClH.H3N/h1H;1H3
InChI key   NLXLAEXVIDQMFP-UHFFFAOYSA-N

Description

Packaging

1 kg in Glass bottle

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store at room temperature. Stock solutions are stable for up to 1 year at room temperature.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
WGK 1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?