• USA Home
  • 178463 - Anti-Apolipoprotein A-I Goat pAb

EMAIL THIS PAGE TO A FRIEND
178463 Sigma-Aldrich

Anti-Apolipoprotein A-I Goat pAb

Anti-Apolipoprotein A-I, goat polyclonal, recognizes apolipoprotein A-I in human. It is validated for Western blotting, immunoprecipitation, immunoelectrophoresis, and paraffin sections.

Purchase

Properties

Related Categories AO-AQ, Alphabetical Index, Antibodies, Primary Antibodies
clone   polyclonal
biological source   goat
species reactivity   human
form   liquid
shipped in   wet ice
isotype   IgG
antibody product type   primary antibodies
Quality Level   100
contains   ≤0.1% sodium azide as preservative
mfr. no.   Calbiochem®
storage condition   OK to freeze
  avoid repeated freeze/thaw cycles
storage conditions   -20C

Description

General description

Goat polyclonal antibody supplied as diluted serum that has been defibrinated, delipidized, and dialyzed against a Tris-HCl buffer. Recognizes the apolipoprotein A-I.

Recognizes apolipoprotein A-I.

  • Antibody Target Gene Symbol: APOA1
    • Target Synonym: Ai, ALP-1, Apo1a, APOA-I, APOLIPOPROTEIN A-I, APOLIPOPROTEIN A-I G1, APOLIPOPROTEIN A1, Brp-14, HDLA1, LP(A-I), Ltw-1, Lvtw-1, MGC102525, MGC117399, Sep-1, Sep-2
    • Entrez Gene Name: apolipoprotein A-I
    • Hu Entrez ID: 335 (Related Antibodies: 178422, 178474, 178472, 178470)
    • Mu Entrez ID: 11806
    • Rat Entrez ID: 25081
  • Immunogen

    Human

    purified human apolipoprotein A-I

    Application

    Immunoblotting (see application references)

    Immunoelectrophoresis (see comments)

    Immunoprecipitation (1.2 mg/ml, Becker titer)

    Paraffin Sections (see application references)

    Physical form

    Serum diluted in 500 mM NaCl, 50 mM Tris-HCl, pH 7.5.

    Reconstitution

    Following initial thaw, aliquot and freeze (-20°C).

    Packaging

    1 ml in Plastic ampoule

    Please refer to vial label for lot-specific concentration.

    Warning

    Toxicity: Standard Handling (A)

    Other Notes

    Monospecific as determined by immunoelectrophoresis (IEP) using 200 µl antibody (as supplied) in 11.8 ml of agar layer, tested against 20 µl of normal human serum. Variables associated with assay conditions will dictate the proper working dilution.

    Safety & Documentation

    Safety Information

    Safety Information for this product is unavailable at this time.

    Documents

    Certificate of Analysis (COA)

    Please Enter a Lot Number
    Protocols & Articles

    Articles

    Antibody Basics

    Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
    Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

    Related Content

    Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

    Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
    Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

    Related Products

    Technical Service:

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Bulk Ordering & Pricing:

    Need larger quantities for your development, manufacturing or research applications?