• USA Home
  • 198901 - Benzalkonium Chloride - CAS 8001-54-5 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
198901 Millipore

Benzalkonium Chloride - CAS 8001-54-5 - Calbiochem

Cationic surface-active agent and germicide. Its effects are potentiated in the presence of di-n-butyl esters, such as diisobutyladipate. M.W. ~360.

Synonym: Benzalkonium Chloride - CAS 8001-54-5 - Calbiochem, Zephiran Chloride

Purchase

Properties

Quality Level   100
form   liquid
mfr. no.   Calbiochem®
storage condition   OK to freeze
storage conditions   +15C to +30C
shipped in   ambient

Description

General description

Cationic surface-active agent and germicide. Its effects are potentiated in the presence of di-n-butyl esters, such as diisobutyladipate.

Cationic surface-active agent and germicide. Its effects are potentiated in the presence of di-n-butyl esters, such as diisobutyladipate.

M.W. ~360

Packaging

250 ml in Metal can

Warning

Toxicity: Corrosive (L)

Physical form

17% aqueous solution.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Gainor, B.J., et al. 1997. J. Orthop. Trauma 11, 121.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?