• USA Home
  • 218742 - Caspase Inhibitor I, Biotin Conjugate - Calbiochem

EMAIL THIS PAGE TO A FRIEND
218742 Sigma-Aldrich

Caspase Inhibitor I, Biotin Conjugate - Calbiochem

The Caspase Inhibitor I, Biotin Conjugate controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym: Biotin-X-VAD-FMK, Caspase Inhibitor I, Biotin Conjugate - Calbiochem

  • Empirical Formula (Hill Notation) C30H49FN6O8S

  • Molecular Weight 672.81

Purchase

Properties

Related Categories A to C, Biochemicals and Reagents, Caspase, Enzyme Inhibitors, Enzyme Inhibitors by Enzyme,
Quality Level   100
form   solid
mfr. no.   Calbiochem®
storage condition   OK to freeze
  desiccated
storage conditions   -20C
color   brown
solubility   DMSO: 20 mM
shipped in   ambient

Description

General description

Biotinylated derivative of Caspase Inhibitor I (Cat. No. 627610) that may be used to identify active caspases in cell lysates.

Biotinylated derivative of Caspase Inhibitor I (Cat. No. 627610) that may be used to identify active caspases in cell lysates.

Packaging

1 mg in Plastic ampoule

Biochem/physiol Actions

Cell permeable: no

Primary Target
Caspase

Product does not compete with ATP.

Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Biotin-X-Val-Ala-Asp(OMe)-CH₂F* (X = linker)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Analysis Note

single spot by TLC

Other Notes

Li, S. et al. 1999. J. Biol. Chem.274, 17325.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?