• USA Home
  • 230746 - Anti-β-Chorionic Gonadotropin Mouse mAb (P3G7)

EMAIL THIS PAGE TO A FRIEND
230746 Sigma-Aldrich

Anti-β-Chorionic Gonadotropin Mouse mAb (P3G7)

liquid, clone P3G7, Calbiochem®

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, B-BI, Primary Antibodies
clone   P3G7, monoclonal
biological source   mouse
species reactivity   human
form   liquid
shipped in   wet ice
isotype   IgG1
Quality Level   100
antibody product type   primary antibodies
contains   ≤0.1% sodium azide as preservative
mfr. no.   Calbiochem®
storage condition   OK to freeze
  avoid repeated freeze/thaw cycles
storage conditions   -20C

Description

General description

Mouse monoclonal antibody purified by DEAE chromatography. Recognizes β-chorionic gonadotropin.

Recognizes β-chorionic gonadotropin.

This Anti-β-Chorionic Gonadotropin Mouse mAb (P3G7) is validated for use in Enzyme Immunoassay, Radioimmunoassay for the detection of β-Chorionic Gonadotropin.

Immunogen

Human

the β chain of hCG

Application

Enzyme Immunoassay (see comments)

Radioimmunoassay (see comments)

Physical form

In 150 mM NaCl, 15 mM potassium phosphate, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Packaging

1 mg in Plastic ampoule

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Specifically designed for EIA and RIA; working dilution should be determined for individual sample types.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?