• USA Home
  • 263301 - Dermatan Sulfate, Porcine Intestinal Mucosa - CAS 54328-33-5 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
263301 Millipore

Dermatan Sulfate, Porcine Intestinal Mucosa - CAS 54328-33-5 - Calbiochem

Mucopolysaccharide containing iduronic acid. Shown to inhibit adenylate kinase from rat hepatic membranes and acts as a potent mediator of fibroblast growth factor-2 responsiveness.

Synonym: Iduronic Acid-β-1,4-N-acetyl-D-galactosamine, Chrondroitin Sulfate B

Purchase

Properties

Related Categories Biochemicals and Reagents, Glycan Component Classes, Glycobiology, Glycosaminoglycans, Glycosaminoglycans and Proteoglycans,
packaging   250 mg in Plastic ampoule (263301-250MG)
color   white
form   solid
shipped in   ambient
solubility   water: 5 mg/mL
storage condition   OK to freeze
brand family   Calbiochem
storage conditions   +2C to +8C

Description

General description

Iduronic acid-containing mucopolysaccharide abundant in skin, arterial walls, and heart valves. Shown to inhibit both Gpp(NH)p- and FSK-stimulated adenylate cyclase activity in rat hepatic membrane preparations and act as an active mediator of fibroblast growth factor-2 responsiveness in wound healing via FGF-2 affinity interaction (KD 2.48 µM). M.W. 30,000 (avg.).

M.W. 30,000 (avg.).

Other Notes

Dittmann, J., et al. 1998. Life Sci. 63, 2199.
Penc, S.F., et al. 1998. J. Biol. Chem. 273, 28116.

Packaging

250 mg in Plastic ampoule

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months ant -20°C.

Warning

Standard Handling (A)

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?