• USA Home
  • 324894 - Epigenetic Regulators Panel - Methylation - Calbiochem

EMAIL THIS PAGE TO A FRIEND
324894 EMD Millipore

Epigenetic Regulators Panel - Methylation - Calbiochem

Purchase

Properties

Related Categories Bioactive Small Molecules, Cell Biology More...
brand family   Calbiochem
packaging   1 ea in Glass bottle (324894-1EA)
form   Solid
toxicity   Regulatory Review
shipped in   wet ice
storage temp.   -20C
storage conditions   Protect from light
  Ok to freeze

Description

Application data

Packaging

Packaged under inert gas

Storage and Shipping Instructions

Following reconstitution, aliquot and freeze (-20°C).

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?