• USA Home
  • 401416 - Goat Anti-Rat IgG, H & L Chain Specific Peroxidase Conjugate

EMAIL THIS PAGE TO A FRIEND
401416 Sigma-Aldrich

Goat Anti-Rat IgG, H & L Chain Specific Peroxidase Conjugate

This Goat Anti-Rat IgG, H & L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Rat IgG, H & L Chain Specific.

Purchase

Properties

Related Categories Antibodies, Secondary Antibodies More...
brand family   Calbiochem
packaging   2 ml in Plastic ampoule (401416-2ML)
form   Liquid
preservative   0.02% Thimerosal
concentration   Please refer to vial label for lot-specific concentration
isotype   IgG
clone   Polyclonal Antibody
host   Goat
toxicity   Standard Handling
shipped in   wet ice
storage temp.   +2C to +8C
storage conditions   Yes

Description

Overview

Immunoaffinity purified goat polyclonal antibody conjugated to horseradish peroxidase (HRP). Recognizes rat IgG, heavy and light chains.

Other Notes

o-Phenylenediamine, 1 mg/ml in citrate buffer, pH 4.5, 0.1% urea peroxide at 25°C, was used as substrate for testing immunoenzyme reactivity. Monospecific for rat IgG, heavy and light chains, as determined by immunoelectrophoresis (IEP) against normal rat serum. Variables associated with assay conditions will dictate the proper working dilution.

Formulation

In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Application

Enzyme Immunoassay (1:10,000)
Immunoelectrophoresis (see comments)

Mercury prohibited

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?