• USA Home
  • 4074 - D-(+)-Glucose monohydrate

EMAIL THIS PAGE TO A FRIEND
4074 Millipore

D-(+)-Glucose monohydrate

Synonym: Dextrose monohydrate

  • CAS Number 14431-43-7

  • Empirical Formula (Hill Notation) C6H12O6 · H2O

  • Molecular Weight 198.17

  •  Beilstein/REAXYS Number 5250278

  •  MDL number MFCD00149450

Purchase

Properties

mfr. no.   Calbiochem®
storage condition   OK to freeze
shipped in   ambient
SMILES string   O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O
InChI   1S/C6H12O6.H2O/c7-1-3(9)5(11)6(12)4(10)2-8;/h1,3-6,8-12H,2H2;1H2/t3-,4+,5+,6+;/m0./s1
InChI key   SPFMQWBKVUQXJV-BTVCFUMJSA-N

Description

General description



Suitable for biochemistry.

Specifications:
Heavy metals (as Pb): 0.001% max
Identity (IR-spectrum): Conforms
Maltose (TLC): 0.2% max
Specific rotation [a]20/D (c=10, water, anhydrous): +52.5-+53.0°
Water (H2O): 10% max





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Packaging

5 kg in Plastic bottle

500 g in Glass bottle

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?