• USA Home
  • 4801 - Neomycin Sulfate - CAS 1405-10-3 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
4801 Sigma-Aldrich

Neomycin Sulfate - CAS 1405-10-3 - Calbiochem

Synonym: Neomycin Sulfate - CAS 1405-10-3 - Calbiochem

  • CAS Number 1405-10-3

  • Empirical Formula (Hill Notation) C23H46N6O13 · 3H2SO4

  • Molecular Weight 908.88

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Enzyme Inhibitors, Enzyme Inhibitors by Enzyme,
Quality Level   200
form   solid
specific activity activity   ≥600 IU/mg
mfr. no.   Calbiochem®
storage condition   OK to freeze
  desiccated
  protect from light
storage conditions   +15C to +30C
color   white
solubility   water: 6 mg/mL
shipped in   ambient
InChI   1S/C23H46N6O13.H2O4S/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;1-5(2,3)4/h5-23,30-36H,1-4,24-29H2;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22?,23+;/m1./s1
InChI key   OIXVKQDWLFHVGR-GQTDVWSESA-N

Description

Packaging

25 g in Plastic ampoule

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Safety & Documentation

Safety Information

Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?