• USA Home
  • 5024 - LIGHT DIAGNOSTICS Goat Anti-Mouse IgG Antibody FITC Reagent, F(ab)′2 (CMV kits only) ~250 tests

EMAIL THIS PAGE TO A FRIEND
5024 Sigma-Aldrich

LIGHT DIAGNOSTICS Goat Anti-Mouse IgG Antibody FITC Reagent, F(ab)′2 (CMV kits only) ~250 tests

from mouse, FITC conjugate

  •  eCl@ss 32161000

Purchase

Properties

Related Categories Antibodies, FITC Antibodies, Mouse Secondary Antibodies and Conjugates, Secondaries listed by Conjugate, Secondaries listed by Target Species,
Quality Level   500
biological source   mouse
antibody product type   secondary antibodies
clone   monoclonal
mfr. no.   Chemicon®
  LIGHT DIAGNOSTICS
application(s)   immunofluorescence: suitable
conjugate   FITC conjugate
detection method   fluorescent
shipped in   wet ice

Description

General description

Diagnostics Kit

Immunogen

Epitope: F(ab)′2 (CMV kits only) ~250 tests

Application

Use undiluted. Refer to Kit Package Insert for directions.

Physical form

Liquid provided as a solution in Phosphate Buffered Saline (PBS), containing 0.02% Evans Blue counterstain, 0.2% BSA, and 0.1% Sodium Azide as a preservative.

Storage and Stability

When stored at 2-8°C and protected from light, this reagent is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures.

Warnings / Precautions:

1) Sodium Azide can form potentially explosive metal azides with plumbing. Flush with large volumes of water to prevent azide build up.

2) Avoid contact with Evans Blue, as it is a potential carcinogen. If skin contact occurs, flush with large volumes of water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For in vitro Diagnostic Use

CE Mark

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?