• USA Home
  • 528875 - PEG 1000 - CAS 25322-68-3 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
528875 Millipore

PEG 1000 - CAS 25322-68-3 - Calbiochem

Linear polymer of ethylene oxide. Melting point 38-42°C.

Synonym: PEG, Poly(ethylene glycol), Polyethylene Glycol 1000

Purchase

Properties

Quality Level   100
form   solid
mfr. no.   Calbiochem®
storage condition   OK to freeze
storage conditions   +15C to +30C
color   white
solubility   water: 10 mg/mL
shipped in   ambient
InChI   1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key   LYCAIKOWRPUZTN-UHFFFAOYSA-N

Description

General description

Linear polymer of ethylene oxide. Melting point 38-42°C.

Water-soluble, linear polymer of ethylene oxide.

Packaging

25 g in Glass bottle

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at -20°C.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?