• USA Home
  • 575303 - T Antigen, p-Nitrophenyl- - CAS 59837-14-8 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
575303 Sigma-Aldrich

T Antigen, p-Nitrophenyl- - CAS 59837-14-8 - Calbiochem

Useful as a substrate for α2,3-(O)-Sialyltransferase.

Synonym: p-Nitrophenyl-2-acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside, Galβ1,3GalNAcα-PNP, T Antigen, p-Nitrophenyl- - CAS 59837-14-8 - Calbiochem

  • CAS Number 59837-14-8

  • Empirical Formula (Hill Notation) C20H28N2O13

  • Molecular Weight 504.44

  •  MDL number MFCD00064051

Purchase

Properties

Quality Level   100
assay   ≥97% (TLC)
form   lyophilized
mfr. no.   Calbiochem®
storage condition   OK to freeze
storage conditions   -20C
solubility   water: 10 mg/mL
  methanol: soluble
shipped in   ambient
InChI   1S/C20H28N2O13/c1-8(25)21-13-18(35-20-17(29)16(28)14(26)11(6-23)34-20)15(27)12(7-24)33-19(13)32-10-4-2-9(3-5-10)22(30)31/h2-5,11-20,23-24,26-29H,6-7H2,1H3,(H,21,25)/t11-,12-,13-,14+,15+,16+,17-,18?,19+,20+/m1/s1
InChI key   INMOOBMAIAWVBW-SLMMFOMASA-N

Description

General description

Useful as a substrate for α2,3-(O)-Sialyltransferase (Cat. No. 566227).

Useful as a substrate for α2,3-(O)-Sialyltransferase (Cat. No. 566227).

Packaging

5 mg in Plastic ampoule

Biochem/physiol Actions

Cell permeable: no

Primary Target
Useful as a substrate for α2,3-(O)-Sialyltransferase

Product does not compete with ATP.

Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?