• USA Home
  • 662091 - Ubiquitin Conjugating Enzyme 5a, GST-Fusion, Human, Recombinant, E. coli

EMAIL THIS PAGE TO A FRIEND
662091 Sigma-Aldrich

Ubiquitin Conjugating Enzyme 5a, GST-Fusion, Human, Recombinant, E. coli

Synonym: GST-Ubc5a (E2)

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Enzymes, Inhibitors, and Substrates, Protein & Nucleic Acid Isolation
assay   ≥95% (SDS-PAGE)
form   liquid
mfr. no.   Calbiochem®
storage condition   OK to freeze
  avoid repeated freeze/thaw cycles
storage conditions   = -70C
shipped in   wet ice

Description

General description

This product has been discontinued.



Recombinant, human ubiquitin conjugating enzyme 5a fused to GST and expressed in E. coli. E2s are a family of ubiquitin carrier proteins that transfer ubiquitin to the ε-lysine of proteins designated for degradation. UbcH5a is a human homolog of the yeast UBC4/5 family which has been shown to play many important regulatory roles in inflammation and cancer.

E2s are a family of ubiquitin carrier proteins that transfer ubiquitin to the ε-lysine of proteins designated for degradation. UbcH5a is a human homolog of the yeast UBC4/5 family that has been shown to play many important regulatory roles in inflammation and cancer.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM HEPES, 50 mM NaCl, 1 mM DTT, 10% glycerol, pH 7.6.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Varshavsky, A. 1997. Trends Biochem. Sci. 22, 383.
Hochstrasser, M. 1996. Annu. Rev. Genet. 30, 405.
Ciechanover, A. 1994. Cell 79, 13.
Seufert, W., and Jentsch, S. 1990. EMBO J. 9, 543.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?