• USA Home
  • 8.17004 - Polyethylene glycol 600

EMAIL THIS PAGE TO A FRIEND
8.17004 SAFC

Polyethylene glycol 600

EMPROVE® ESSENTIAL Ph Eur

Synonym: PEG, Poly(ethylene glycol), Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 600

Purchase

Properties

Agency/Method   Ph Eur
Quality Level   500
vapor pressure   <0.1 hPa ( 20 °C)
product line   Emprove® Essential
autoignition temp.   >320 °C (DIN 51794)
potency   28000 mg/kg LD50, oral (Rat)
  >20000 mg/kg LD50, skin (Rabbit)
pH   5-7 (20 °C, 100 g/L in H2O)
mp   17-22 °C
transition temp   flash point 260 °C
density   1.13 g/cm3 at 20 °C
SMILES string   C(CO)O
InChI   1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key   LYCAIKOWRPUZTN-UHFFFAOYSA-N
storage conditions   no restrictions.

Description

Analysis Note

Identity: passes test
Appearance: clear, viscous, colourless or almost colourless hygroscopic liquid
Appearance of solution: passes test
Acidity,Alkalinity: passes test
Reducing substances: passes test
Density (d 20/20): about 1.080 (50% solution)
Solidification point: 15 - 25 °C
Viscosity (20 °C): 14.8 - 17.6 mm²/s
Viskosity (20 °C): 16.0 - 19.0 mPa∙s
Formaldehyde: ≤ 30 ppm
Ethylene glycol and diethylene glycol: ≤ 0.4 %
Ethylene oxide (GC): ≤ 1 ppm
Dioxane (GC): ≤ 10 ppm
Hydroxyl value: 178 - 197
Average molecular mass: 570 - 630
Sulfated ash: ≤ 0.2 %
Water (K. F.): ≤ 2.0 %
Other residual solvents acc. to ICH-guideline are excluded by the manufacturing process.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not
likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Emprove® Dossiers

Already an EMPROVE® Program customer? Sign in to access your EMPROVE® Dossiers

Sign In
What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Three document types are included with an Emprove® Program subscription:

  • Material Qualification Dossiers
  • Quality Management Dossiers
  • Operational Excellence Dossiers

Learn more about the benefits an Emprove® Program subscription can provide

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?