EMAIL THIS PAGE TO A FRIEND
8.45018 Sigma-Aldrich

Ethylenediamine

Msynthplus

Synonym: 1,2-Diaminoethane, 1,2-Ethanediamine, 1,2-Diaminoethane

  • CAS Number 107-15-3

  • Linear Formula NH2CH2CH2NH2

  • Molecular Weight 60.10

  •  Beilstein/REAXYS Number 605263

  •  MDL number MFCD00008204

  •  EC Index Number 203-468-6

Purchase

Properties

Quality Level   400
vapor density   2.07 (vs air)
vapor pressure   10 mmHg ( 20 °C)
assay   ≥99% (GC)
form   liquid
autoignition temp.   716 °F
potency   866 mg/kg LD50, oral (Rat)
  560 mg/kg LD50, skin (Rabbit)
expl. lim.   16 %
refractive index   n20/D 1.4565 (lit.)
pH   12.2 (20 °C, 100 g/L in H2O)
bp   118 °C (lit.)
mp   8.5 °C (lit.)
transition temp   flash point 38 °C
density   0.899 g/mL at 25 °C (lit.)
SMILES string   NCCN
InChI   1S/C2H8N2/c3-1-2-4/h1-4H2
InChI key   PIICEJLVQHRZGT-UHFFFAOYSA-N
storage conditions   Store below +30°C.

Description

Analysis Note

Color (visual): colourless
Appearance of substance (visual): clear
Assay (GC, area%): ≥ 99.0 % (a/a)
Water (GC, area%): ≤ 1.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.896 - 0.898
Identity (IR): passes test
1,1-Dichloroethene (HS-GC): lot specific result
1,1,1-Trichloroethane (HS-GC): lot specific result
Benzene (HS-GC): lot specific result
Carbon tetrachloride (HS-GC): lot specific result
1,2-Dichlorethane: not measurable

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?