• USA Home
  • 8.45160 - 1-Bromo-4-chlorobutane

EMAIL THIS PAGE TO A FRIEND
8.45160 Sigma-Aldrich

1-Bromo-4-chlorobutane

Msynthplus

Synonym: 1-Bromo-4-chlorobutane, Tetramethylene chlorobromide

Purchase

Properties

Quality Level   400
bp   175-176 °C/1013 hPa
transition temp   flash point 60 °C
density   1.489 g/cm3 at 20 °C
InChI   1S/C4H8BrCl/c5-3-1-2-4-6/h1-4H2
InChI key   NIDSRGCVYOEDFW-UHFFFAOYSA-N
storage conditions   Store below +30°C.

Description

General description

Msynthplus

Application

1-Bromo-4-chlorobutane Msynthplus. CAS 6940-78-9, molar mass 171.46 g/mol.

Analysis Note

Colour (visual)colourlessAppearance of substance (visual)clearAssay (GC, area%)≥ 97.0 % (a/a)Density (d 20 °C/ 4 °C)1.487 - 1.494Identity (IR)passes test

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?