• USA Home
  • 9290-OP - OmniPur TRIS Solution 1.0M pH 8.0 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
9290-OP Millipore

OmniPur TRIS Solution 1.0M pH 8.0 - Calbiochem

Purchase

Properties

Related Categories Loading Buffer and Tracking Dyes, Molecular Biology, Nucleic Acid Electrophoresis More...
Quality Level   100
form   liquid
mfr. no.   Calbiochem®
color   clear colorless
shipped in   ambient

Description

General description



Molecular biology grade.

Specifications:
DNase: None detected
Concentration (molarity): 0.9-1.1 M
pH of solution at 25°C: 7.9-8.1
Protease: None detected
RNase: None detected
Sterility: To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Buffers - From Bnech to Bulk
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?