• USA Home
  • GF073 - Stromal Cell-Derived Factor-1α Protein, Recombinant human

EMAIL THIS PAGE TO A FRIEND
GF073 Sigma-Aldrich

Stromal Cell-Derived Factor-1α Protein, Recombinant human

The Stromal Cell-Derived Factor-1alpha protein (or SDF1alpha protein) is a recently discovered protein belonging to the alpha-chemokine (C-X-C) family of cytokines.

Synonym: SDF-1alpha

  •  eCl@ss 32160405

Purchase

Properties

biological source   human
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −20°C
Quality Level   100
assay   >98% (SDS-PAGE and HPLC)
mfr. no.   Chemicon®
input   sample type hematopoietic stem cell(s)
NCBI accession no.   NM_000609.4
  NM_001033886.1
  NM_199168.2
UniProt accession no.   P48061
Gene Information   human ... CXCL12(6387)

Description

General description

Stromal Cell-Derived Factor-1alpha (SDF1alpha) is a recently discovered protein belonging to the alpha-chemokine (C-X-C) family of cytokines. GF073 is a 8.0 kDa protein containing 68 amino acid residues.

Storage and Stability

The lyophilized powder, though stable at room temperature, is best stored desiccated below 0°C. Reconstitute in water to a concentration of 0.1-1 mg/mL. It is recommended that further dilutions be made into buffer containing carrier protein or medium containing serum. Reconstituted SDF1alpha should be stored at 4°C or in undiluted aliquots at -20°C for up to 6 months.

Analysis Note

Specific Activity: Determined by its ability to chemoattract human peripheral T cells activated with PHA and IL-2 using a concentration range of 20.0-80.0 ng/mL.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?