• USA Home
  • IL008 - Interleukin-8 Protein, Recombinant human, (72 amino acid form)

EMAIL THIS PAGE TO A FRIEND
IL008 Sigma-Aldrich

Interleukin-8 Protein, Recombinant human, (72 amino acid form)

The recombinant Human interleukin-8 protein (or IL-8 protein) is an 8.4 kDa protein containing 72 amino acid residues.

Synonym: IL-8

  •  eCl@ss 32160405

Purchase

Properties

Quality Level   100
biological source   human
assay   >98% (SDS-PAGE and HPLC)
mfr. no.   Chemicon®
application(s)   cell analysis: suitable (cell culture)
input   sample type hematopoietic stem cell(s)
NCBI accession no.   NM_000584.2
UniProt accession no.   P10145
shipped in   dry ice

Description

General description

Human interleukin-8 (IL-8) is an 8.4 kDa protein containing 72 amino acid residues. It promotes neutrophil chemotaxis and degranulation. The 72 amino acid IL-8 is the predominant form secreted by monocytes and lymphocytes.

Biochem/physiol Actions

Human IL-8 produces chemotaxis over a wide concentration range in an assay using human peripheral blood neutrophils. Significant chemotaxis was achieved using a concentration of 10-100 ng/mL.

Storage and Stability

The lyophilized powder, though stable at room temperature, is best stored desiccated below 0°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can be diluted into other buffered solutions. Reconstituted IL-8 should be stored in undiluted aliquots at -20°C for up to six months.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?