• USA Home
  • MXRPSPRKT - Milliflex Rapid AutoSpray Station

EMAIL THIS PAGE TO A FRIEND
MXRPSPRKT Millipore

Milliflex Rapid AutoSpray Station

Rapid detection, response and resolution of microbial contamination

  •  eCl@ss 32160606

  •  NACRES NB.79

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Millipore - Manufacturer Browser, Millipore Filters, Millipore Filtration Vacuum,
AC/DC input   110 V
  230 V
Quality Level   100
material   316L stainless steel holder (for membrane)
  316L stainless steel holder (for pad)
  316L stainless steel turntable(s)
  polyester keys
  polyurethane case
  polyurethane cover
mfr. no.   Milliflex®
parameter   15-40  °C temperature
H   31 cm (12 in.)
W   21 cm (8.3 in.)
depth   40 cm (101.6 in.)
weight   10 kg (22 lb)
input   sample type pharmaceutical(s)
Featured Industry   Industrial QC
Pharmaceutical
detection method   bioluminescent
shipped in   ambient

Description

General description

An integral component in the Milliflex Rapid Microbiology Detection and Enumeration system, the Milliflex Rapid AutoSpray Station applies an equal volume of two reagents across a membrane surface in order to detect living microorganisms. Using a proven, pre-set automated program, the AutoSpray Station ensures reproducible results, test after test. Reagent application is achieved using a back and forth sweeping movement of the filter holder under the calibrated glass nebulizer. In automatic spray mode, the AutoSpray Station delivers both reagents in 90 seconds at the touch of a button.
From the control panel, analysts can easily initiate an operation by selecting from a list of operations.
Pinch valves ensure that the same volume of reagent is dispensed during each cycle. An internal compressor allows stand-alone operation, eliminating the need for additional equipment.

Application

Type of Test
Bioluminescence

Components

Includes: AutoSpray station

Filter holder

Pad holder

Nebulizer cleaner

5ml female syringe

3nebulizers

silicone liquid tubes

2stainless steel caps

Universal power supply adaptor

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?