• USA Home
  • R03203 - Albumin-Glycerol Fixative

EMAIL THIS PAGE TO A FRIEND
R03203 Sigma-Aldrich

Albumin-Glycerol Fixative

HARLECO® Mayers`s fixative

Synonym: Mayers`s fixative

Purchase

Properties

Quality Level   300
application(s)   hematology: suitable
  histology: suitable
transition temp   flash point 160 °C
Featured Industry   Diagnostic Assay Manufacturing
shipped in   ambient
storage conditions   No declaration

Description

Packaging

100, 500 ml in Plastic bottle

Application

HARLECO® Mayers`s fixative

Used for Mounting Tissue Sections

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?