• USA Home
  • SCGVU02RE - Stericup-GV Sterile Vacuum Filtration System

EMAIL THIS PAGE TO A FRIEND
SCGVU02RE Millipore

Stericup-GV Sterile Vacuum Filtration System

250 mL process volume, 0.22 µm pore size, PVDF membrane, pkg of 12, radio-sterilized

  •  eCl@ss 32031623

  •  NACRES NB.22

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Millipore - Manufacturer Browser, Millipore Filters, Millipore Filtration Vacuum,
Quality Level   400
material   PVDF membrane
  polystyrene
sterility   sterile
feature   holdup volume 2.4 mL
  hydrophilic
mfr. no.   Stericup®
parameter   45 °C max. temp.
application(s)   sterile filtration: suitable
H   164 mm
diam.   12.5 cm
filter diam.   73 mm
filtration area   40 cm2
funnel capacity   250 mL
matrix   Durapore®
pore size   0.22 μm nominal pore size
  0.22 μm pore size
test parameters   sample volume: 250 mL
fitting   inlet funnel
  outlet flask (Receiver)
shipped in   ambient

Description

General description

250 mL bottle with cap

The Stericup® vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format. Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device. The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications

Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component

Application

250 mL process volume, 0.22 µm pore size, PVDF membrane, pkg of 12, radio-sterilized

Principle

Vacuum

Physical form

Color Code: Clear with blue adapter

Preparation Note

Sterilization Method
Gamma irradiation

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany

Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?