• USA Home
  • SCHVU01RE - Stericup-HV Sterile Vacuum Filtration System

EMAIL THIS PAGE TO A FRIEND
SCHVU01RE Millipore

Stericup-HV Sterile Vacuum Filtration System

150 mL process volume, 0.45 µm pore size, PVDF membrane, pkg of 12, radio-sterilized

  •  eCl@ss 32031623

  •  NACRES NB.22

Purchase

Properties

packaging   12 (SCHVU01RE)
H   120 mm
application(s)   sterile filtration: suitable
diam.   12.5 cm
feature   holdup volume 2.4 mL
  hydrophilic
filter diam.   73 mm
filtration area   40 cm2
fitting   inlet funnel
  outlet flask (Receiver)
funnel capacity   150 mL
material   PVDF membrane
  polystyrene
matrix   Durapore
mfr. no.   Stericup
parameter   150 mL process volume
  45 °C max. temp.
pore size   0.45 μm nominal pore size
  0.45 μm pore size
shipped in   ambient
test parameters   sample volume: 150 mL
sterility   sterile

Description

Application

150 mL process volume, 0.45 µm pore size, PVDF membrane, pkg of 12, radio-sterilized

General description

150 mL bottle with cap

The Stericup® vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format. Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device. The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications

Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component

Principle

Vacuum

Preparation Note

Sterilization Method
Gamma irradiation

Physical form

Color Code: Clear with blue adapter

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?