• USA Home
  • SCHVU11RE - Stericup-HV Sterile Vacuum Filtration System

EMAIL THIS PAGE TO A FRIEND
SCHVU11RE Millipore

Stericup-HV Sterile Vacuum Filtration System

1000 mL process volume, 0.45 µm pore size, PVDF membrane, pkg of 12, radio-sterilized

Purchase

Properties

trade name   Stericup
fitting inlet   Funnel
fitting outlet   Receiver Flask
feature   1000 mL bottle with cap
maximum operating temperature   45 °C
pressure/vacuum   Vacuum
application(s)   Sterile Filtration
media   Durapore®
sterility   Sterile
sterilization   Gamma irradiation
wettability   Hydrophilic
hold-up volume   2.4 mL
pore size   0.45 µm
nominal pore size   0.45 µm
height   319 mm
funnel capacity   1 L
diameter   13.5 cm
filtration area   40 cm²
volume   1 L
filter diameter (ø)   73 mm
process volume   1000 mL
chemistry   Hydrophilic Polyvinylidene Fluoride (PVDF)
device material   Polystyrene
material size   12
shipped in   ambient
color code   Clear with blue adapter

Description

Background information

The Stericup® vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop® filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format. Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device. The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop® bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

Millipore Express® PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore® membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup® device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications

Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?