• USA Home
  • AB3256-100UL - Anti-Luciferase Antibody, Firefly

EMAIL THIS PAGE TO A FRIEND
AB3256-100UL Sigma-Aldrich

Anti-Luciferase Antibody, Firefly

Anti-Luciferase Antibody, Firefly detects level of Luciferase & has been published & validated for use in ELISA, IP, WB, IH.

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
trade name   Chemicon
brand family   Chemicon
format   Purified
application(s)   ELISA
  Immunoprecipitation
  Western Blotting
  Immunohistochemistry
clone   Polyclonal Antibody
epitope   Firefly
host   Goat
species reactivity   Invertebrates
UniProt accession no.   Q27321
material size   100 µL
shipped in   wet ice
storage conditions   Maintain frozen at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Description

Physical form

Immunoglobulin purified by delipidation, salt fractionation and ion exchange chromatography. Liquid in 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% sodium azide.

Application

ELISA: 1:50,000-1:200,000 when assayed against 1 μg of Luciferase in a standard sandwich ELISA.

Immunoblotting: 1:5,000-1:20,000

Immunohistochemistry: 1:1,000-1:5,000

Immunoprecipitation

Optimal working dilutions must be determined by end user.

Specificity

Luciferase [Photinus pyralis (Firefly)]. Monospecific by IEP. May show cross reactivity with Luciferase from other sources.

Immunogen

Luciferase [Photinus pyralis (Firefly)].

Usage Statement

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?