EMAIL THIS PAGE TO A FRIEND
ABN335 Sigma-Aldrich

Anti-LGI2 Antibody

Detect the LGI2 protein using this Anti-LGI2 validated for use in WB.

Synonym: Leucine-rich, glioma inactivated 2, LGIL2

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
packaging   100 µg (ABN335)
application(s)   western blot: suitable
biological source   rabbit
clone   polyclonal
purified by   affinity chromatography
shipped in   wet ice
species reactivity   human
gene symbol   LGI2(55203)
molecular weight   ~63 kDa observed. An uncharacterized band may appear at ~49 kDa in some lysates.
concentration   Please refer to the Certificate of Analysis for the lot-specific concentration.
gene symbol   Lgi2
Quality Assurance   Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected LGI2 in 15 µg/mL of HEK293 cell lysate.

Description

Target description

~63 kDa observed. An uncharacterized band may appear at ~49 kDa in some lysates.

Application

Detect the LGI2 protein using this Anti-LGI2 validated for use in WB.

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected LGI2 in human midbrain tissue.

General description

Leucine-rich repeat LGI family member 2 (LGI2) is a member of the LGI family. The LGI family consists of four highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI2 have not been found to be associated with ADTLE.

Immunogen

KLH-conjugated linear peptide corresponding to the N-terminus of human LGI2.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Specificity

This LGI2 antibody is predicted to be specific to LGI2 and not recognize other LGI proteins. Three isoforms of LGI2 are known to exist; this LGI2 antibody will recognize only the longest.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Making Headway
Protocols & Articles

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?