• USA Home
  • ABN485 - Anti-SIPA1L1 Antibody

EMAIL THIS PAGE TO A FRIEND
ABN485 Sigma-Aldrich

Anti-SIPA1L1 Antibody

Anti-SIPA1L1 Antibody is a highly specific rabbit polyclonal antibody, that targets Signal-induced proliferation associated-like protein 1 & has been tested in western blotting & IHC.

Synonym: Signal-induced proliferation associated-like protein 1, SIPA1-like protein 1, E6TP1, High-risk human papilloma viruses E6 oncoproteins targeted protein 1, E6-targeted protein 1, SIPA1L1

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, SG-SK
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry: suitable
  western blot: suitable
species reactivity   human, rat
shipped in   wet ice
Quality Level   100
antibody product type   primary antibodies
purified by   affinity chromatography
UniProt accession no.   O43166
Gene Information   human ... SIPA1L1(26037)

Description

General description

Signal-induced proliferation associated-like protein 1 (SIPA1L1), also called High-risk human papilloma viruses E6 oncoproteins targeted protein 1 (E6-targeted protein 1) is widely expressed and bound at the carboxy-terminus by Casein kinase I epsilon (CKIe) which leads to phosphorylation. SIPA1L1 activates RAP2A GTPase activity, aids reorganization of the actin cytoskeleton, recruits DLG4 to F-actin, regulates dendritic spine morphogenesis, and plays important roles in embryo development and cell proliferation.

Immunogen

KLH-conjugated linear peptide corresponding to human SIPA1L1.

Application

Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected SIPA1L1 in human brain tissue.

Target description

~200 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 2 isoforms at ~198 kDa and ~200 kDa

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected SIPA1L1 in 15µg of rat brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?