• USA Home
  • AP1155 - Anti-PON1 Mouse mAb (2H7)

EMAIL THIS PAGE TO A FRIEND
AP1155 Sigma-Aldrich

Anti-PON1 Mouse mAb (2H7)

liquid, clone 2H7, Calbiochem®

Synonym: Anti-Paraoxonase 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, POD-POZ, Primary Antibodies
clone   2H7, monoclonal
biological source   mouse
species reactivity   human
form   liquid
shipped in   wet ice
isotype   IgG2a
Quality Level   100
antibody product type   primary antibodies
does not contain   preservative
mfr. no.   Calbiochem®
storage condition   OK to freeze
  avoid repeated freeze/thaw cycles
storage conditions   -20C

Description

General description

Purified mouse monoclonal antibody. Recognizes the PON1 protein.

Recognizes the PON1 protein.

This Anti-PON1 Mouse mAb (2H7) is validated for use in ELISA, Immunoblotting for the detection of PON1.

Immunogen

A recombinant polypeptide corresponding to amino acids 246-356 of human PON1, expressed as a GST fusion protein

Application


ELISA (see comments)

Immunoblotting (1-5 µg/ml)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Warning

Toxicity: Regulatory Review (Z)

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Safety & Documentation

Safety Information

WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?