• USA Home
  • AX0612 - Alumina Chromatographic Grade

EMAIL THIS PAGE TO A FRIEND
AX0612 Supelco

Alumina Chromatographic Grade

80-200 mesh, Meets Reagent Specifications for testing USP/NF monographs

Synonym: Alumina Chromatographic Grade, Aluminum Oxide

  • CAS Number 1344-28-1

  • Empirical Formula (Hill Notation) Al2O3

  • Molecular Weight 101.96

  •  MDL number MFCD00003424

  •  EC Index Number 215-691-6

Purchase

Properties

Quality Level   200
form   powder
bp   2980 °C/1013 hPa
mp   2050 °C
shipped in   ambient
storage conditions   Store at room temperature.

Description

Packaging

500 g in Glass bottle

Analysis Note

Adsorptive power: 0.2mg/g min
Color: White
Form: Powder
Mesh size (80-200): 70% min
Water (at time of packaging): 10% max
pH (10% suspension in water): 9-10

Safety & Documentation

Safety Information

WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?