• USA Home
  • DX0101 - 1-Decanesulfonic Acid, Sodium Salt, Anhydrous

EMAIL THIS PAGE TO A FRIEND
DX0101 Sigma-Aldrich

1-Decanesulfonic Acid, Sodium Salt, Anhydrous

HPLC

Synonym: 1-Decanesulfonic Acid, Sodium Salt, Anhydrous, Decane Sulfuric Acid

  • CAS Number 13419-61-9

  • Linear Formula CH3(CH2)9SO3Na

  •  Colour Index Number 45400

  •  MDL number MFCD00007526

  •  EC Index Number 236-525-9

Purchase

Properties

mp   >300 °C
shipped in   ambient
InChI   1S/C10H22O3S.Na/c1-2-3-4-5-6-7-8-9-10-14(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1
InChI key   AIMUHNZKNFEZSN-UHFFFAOYSA-M
storage conditions   Store at room temperature.

Description

Packaging

100 g in Plastic bottle

Application

HPLC

Ion Pair Reagent

Analysis Note

Color: White
Form: Crystalline solid
Loss on drying: 1% max
Solubility: To pass test
UV Abs. at 210 nm: 0.5AU max
UV Abs. at 220 nm: 0.1AU max
UV Abs. at 254 nm: 0.04AU max
UV Abs. at 280 nm: 0.04AU max

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?