EMAIL THIS PAGE TO A FRIEND
DX0685 Supelco

1,2-Dichlorobenzene

Synonym: 1,2-Dichlorobenzene, o-Dichlorobenzene, 1

  • CAS Number 95-50-1

  • Linear Formula 1,2-C6H4Cl2

  •  MDL number MFCD00000535

  •  EC Index Number 202-425-9

Purchase

Properties

Quality Level   200
vapor pressure   1.33 hPa ( 20 °C)
assay   ≥98% (GC)
form   liquid
bp   180 °C/1013 hPa
mp   -17 °C
transition temp   flash point 66 °C
density   1.31 g/cm3 at 20 °C
shipped in   ambient
InChI   1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H
InChI key   RFFLAFLAYFXFSW-UHFFFAOYSA-N
storage conditions   Store at room temperature.

Description

Packaging

4 l in Glas bottle pre packed

Analysis Note

Assay (GC): 98% min
Color (APHA): 20 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Target organs 
Respiratory system
WGK Germany 
WGK 2
Flash Point(F) 
150.8 °F - closed cup
Flash Point(C) 
66.0 °C - closed cup
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?