• USA Home
  • FX0282 - Florisil® Adsorbent PR 60-100 Mesh

EMAIL THIS PAGE TO A FRIEND
FX0282 Supelco

Florisil® Adsorbent PR 60-100 Mesh

Meets Reagent Specifications for testing USP/NF monographs

Synonym: Florisil® Adsorbent PR 60-100 Mesh, Magnesium Silicate Powder

  • CAS Number 1343-88-0

  •  EC Index Number 215-681-1

Purchase

Properties

pH   9.5 (20 °C, 100 g/L in H2O, slurry)
shipped in   ambient
storage conditions   Store at room temperature.

Description

Packaging

1 kg in Glas bottle pre packed

Analysis Note

Color: White
Description: To pass test
Form: Powder
Mesh size (60-100): 80% min

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?