EMAIL THIS PAGE TO A FRIEND
HX0304 Supelco

Hexane, 95%

anhydrous, septum-sealed bottle DriSolv

Synonym: n-Hexane, Hexane, anhydrous

  • CAS Number 110-54-3

  • Linear Formula CH3(CH2)4CH3

  • Molecular Weight 86.18

  •  Beilstein/REAXYS Number 1730733

  •  MDL number MFCD00009520

  •  EC Index Number 203-777-6

Purchase

Properties

Quality Level   200
vapor density   ~3 (vs air)
vapor pressure   256 mmHg ( 37.7 °C)
  5.2 psi ( 37.7 °C)
  ~132 mmHg ( 20 °C)
autoignition temp.   453 °F
expl. lim.   7.7 %
refractive index   n20/D 1.375 (lit.)
bp   69 °C (lit.)
mp   −95 °C (lit.)
transition temp   flash point -22 °C
solubility   0.0095 g/L
density   0.659 g/mL at 25 °C (lit.)
shipped in   ambient
SMILES string   CCCCCC
InChI   1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
InChI key   VLKZOEOYAKHREP-UHFFFAOYSA-N
storage conditions   Store at room temperature.

Description

Packaging

1 l in Glas bottle pre packed

100 ml in Glass bottle

Application

Packed under Nitrogen and sealed with a septum.

anhydrous, septum-sealed bottle DriSolv

Analysis Note

AppearanceClear liquid, free from particulatesAssay (n-hexane)95.0% minColor (APHA)10 maxEvaporation residue3ppm maxFiltered through 0.2 µm filterTo pass testWater (H2O)50ppm max

Safety & Documentation

Safety Information

Signal word 
Danger
Target organs 
Central nervous system, Nervous system
WGK Germany 
WGK 2
Flash Point(F) 
-7.6 °F
Flash Point(C) 
-22 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?