• USA Home
  • MAB1913 - Anti-Procollagen Type I Antibody, CT, clone PCIDG10

EMAIL THIS PAGE TO A FRIEND
MAB1913 Sigma-Aldrich

Anti-Procollagen Type I Antibody, CT, clone PCIDG10

ascites fluid, clone PCIDG10, Chemicon®

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PR-PR, Primary Antibodies
clone   PCIDG10, monoclonal
biological source   mouse
application(s)   immunohistochemistry: suitable
species reactivity   guinea pig, bovine, human
should not react with   mouse, hamster, rat
shipped in   dry ice
antibody form   ascites fluid
isotype   IgG1
Quality Level   100
antibody product type   primary antibodies
mfr. no.   Chemicon®
NCBI accession no.   NM_000088.3
UniProt accession no.   P02452
Gene Information   human ... COL1A1(1277)

Description

Specificity

Labels carboxy-terminal pro-peptide of collagen type I. Does not stain mature collagen fibers in tissue, but rather is localized intracellularly in cells producing pro-collagen I. Positive reaction: human, guinea pig, bovine. Negative reaction: mouse, rat, hamster.

Immunogen

Epitope: C-terminus

Human pro-collagen I.

Application

Immunohistochemistry on paraffin embedded tissue sections fixed in acid ETOH, not traditional formalin. (1% HOAC in 95% ETOH) fixation. Suggested starting dilution 1:100 (10-25 μg/mL).

Optimal working dilutions must be determined by end user.

Research Category
Cell Structure

Research Sub Category
ECM Proteins

This Anti-Procollagen Type I Antibody, C-terminus, clone PCIDG10 is validated for use in IH for the detection of Procollagen Type I.

Physical form

Liquid. No preservative.

Storage and Stability

Maintain at -20°C in aliquots up to 12 months. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?