• USA Home
  • MAB8787 - Anti-SARS Antibody, M Specific, clone 19Cd

EMAIL THIS PAGE TO A FRIEND
MAB8787 Sigma-Aldrich

Anti-SARS Antibody, M Specific, clone 19Cd

Anti-SARS Antibody, M Specific, clone 19Cd detects level of SARS & has been published & validated for use in ELISA, IF, WB, NEUT.

  •  eCl@ss 32160702

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, S1-SD
clone   19Cd, monoclonal
biological source   mouse
application(s)   ELISA: suitable
  immunofluorescence: suitable
  neutralization: suitable
  western blot: suitable
species reactivity   human
shipped in   wet ice
isotype   IgM
Quality Level   300
antibody product type   primary antibodies
mfr. no.   Chemicon®

Description

General description

Severe acute respiratory syndrome (SARS) is a viral respiratory illness caused by a coronavirus, called SARS-associated coronavirus (SARS-CoV). Human coronaviruses (HCoVs) were previously only associated with mild diseases. The SARS-CoV genome contains five major open reading frames (ORFs) that encode the replicase polyprotein; the spike (S), envelope (E), and membrane (M) glycoproteins; and the nucleocapsid protein (N).

Specificity

SARS-associated coronavirus (SARS-CoV) M proteins.

Immunogen

Epitope: M Specific

Application

Immunofluorescence

ELISA

Western Blot

Immunoblot

Optimal dilutions must be determined by the end user.

Research Category
Infectious Diseases

Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified

Purified immunoglobulin. Liquid in 0.02M PB, 0.25M sodium chloride with 0.1% sodium azide as a preservative

Storage and Stability

Store at 2 to 8°C for up to 12 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Coronavirus COVID-19 (SARS-CoV-2) Detection, Characterization & Vaccine Production

As a global life science tools and equipment supplier, we are here to help in the response to COVID-19. From supplying raw materials and research products for detection and characterization to produc...
Keywords: PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?