• USA Home
  • MABF147C - Anti-CD45.2 (Mouse), redFluor® 710, clone 104 Antibody

EMAIL THIS PAGE TO A FRIEND
MABF147C Sigma-Aldrich

Anti-CD45.2 (Mouse), redFluor® 710, clone 104 Antibody

Synonym: Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen (L-CA), Lymphocyte antigen 5 (Ly-5), T200, CD45, CD45.2

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
application(s)   flow cytometry: suitable
biological source   mouse
clone   104, monoclonal
species reactivity   mouse
clone   Monoclonal Antibody
concentration   Please refer to lot specific datasheet.
gene symbol   Ly-5
  Ptprc
conjugate   redFluor 710
Quality Assurance   This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.25 μg from a representative lot detected CD45.2 in one million C57Bl/6 splenocytes.

Description

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

Research Category
Inflammation & Immunology

Research Sub Category
Immunoglobulins & Immunology

General description

The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.

Immunogen

Immunogen information for this antibody is proprietary information.

Other Notes

Concentration: Please refer to lot specific datasheet.

Physical form

Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.

Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?