• USA Home
  • MABF355 - Anti-CD45RO (human) Antibody, APC, clone UCHL1

EMAIL THIS PAGE TO A FRIEND
MABF355 Sigma-Aldrich

Anti-CD45RO (human) Antibody, APC, clone UCHL1

Detect CD45RO using this mouse monoclonal antibody, Anti-CD45RO (human) Antibody, APC, clone UCHL1 validated for use in Flow Cytometry, western blotting, IHC & IP.

Synonym: Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, T200, CD45, CD45RO

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
packaging   100 tests (MABF355)
application(s)   flow cytometry: suitable
  immunohistochemistry: suitable
  immunoprecipitation (IP): suitable
  western blot: suitable
biological source   mouse
clone   UCHL1, monoclonal
isotype   IgG2aκ
shipped in   wet ice
species reactivity   human
gene symbol   PTPRC(5788)
molecular weight   147 kDa calculated
clone   Monoclonal Antibody
concentration   Please refer to the Certificate of Analysis for the lot-specific concentration.
gene symbol   CD45
conjugate   Allophycocyanin conjugate
Quality Assurance   The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.

Flow Cytometry Analysis: 5 μL of this antibody detected CD45RO in one million human peripheral blood lymphocytes.

Description

Target description

147 kDa calculated

Application

Western Blotting Analysis: A representative lot detected human CD45RO in Western Blotting. (Ishii T, et al. 2001.)

Immunohistochemistry Analysis: A representative lot detected human CD45RO in Immunohistochemistry. (Yamada M, et al.1996.;Sakkas LI, et al. 1998.)

Immunoprecipitation Analysis: A representative lot detected human CD45RO in Immunoprecipitation. (Ponsford M, et al. 2001.)





General description

Receptor-type tyrosine-protein phosphatase C, also known as Leukocyte common antigen (L-CA) or CD45 and encoded by the human gene PTPRC or CD45 is a transmembrane glycoprotein that belongs to the PTP family. PTPs are signaling molecules that regulate cellular process like cell growth, differentiation and mitotic cycle. CD45 has many isoforms that are distributed throughout the immune system according to cell type. These arise because of alternative splicing of RA, RB, and RC exons. CD45 and its isoforms non-covalently associate with lymphocyte phosphatase-associated phosphoprotein (LPAP) on T and B lymphocytes. CD45RO is one of the isoforms of CD45 that facilitates T cell activation. It is also the shortest isform which lack the RA, RB, and RC exons. Mutations in CD45 have been associated with mulitple sclerosis and severe combined immunodeficiency.



Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?