• USA Home
  • MABF385 - Anti-CD127 (IL-7Ra) Antibody (human), APC, clone R34-34

EMAIL THIS PAGE TO A FRIEND
MABF385 Sigma-Aldrich

Anti-CD127 (IL-7Ra) Antibody (human), APC, clone R34-34

clone R34-34, from mouse

Synonym: Interleukin-7 receptor subunit alpha, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL-7R-alpha, IL-7RA, CDw127, CD antigen CD127

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CD0-CDC, Primary Antibodies
conjugate   Allophycocyanin conjugate
clone   R34-34, monoclonal
biological source   mouse
application(s)   flow cytometry: suitable
species reactivity   human
shipped in   wet ice
packaging   pkg of 5 μL (0.5 μg/test)
isotype   IgG1κ
Quality Level   100
antibody product type   primary antibodies
UniProt accession no.   P16871
Gene Information   human ... IL7R(3575)

Description

General description

The R34-34 antibody is specific for human CD127, a cell surface protein also known as the Interleukin-7 Receptor alpha chain, or IL-7R alpha. CD127 is typically expressed at the cell surface as a heterodimer with the common gamma chain (CD132). This complex acts as the functional receptor for IL-7, a cytokine important in T and B cell development, and in mature T cell homeostasis. A second cytokine known as Thymic Stromal Lymphopoietin (TSLP) also binds to a receptor complex of CD127 and the TSLPR chain to trigger activation of dendritic cells, and is involved in B cell development, allergy and autoimmunity. The R34-34 antibody may be used as a phenotypic marker for CD127 on immature B cells, on subsets of thymocytes which are double negative (CD4-CD8- ) or single positive (CD4+ or CD8+), and at low levels on mature, peripheral T cells. CD127 is a key marker, when used in combination with CD4 and CD25, to distinguish Treg and effector/memory Treg populations known as T(REM), whereas certain populations of Foxp3+ /CD25+ T cells lacking CD127 have been associated with impaired suppressive function (Bikker et al. 2012. Rheumatology, 51: 996-1005)

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology

This Mouse Monoclonal Antibody Anti-CD127 (IL-7Ra) (human), APC, clone R34-34, Cat. No. MABF385 is validated for use in FC for the detection of CD127 (IL-7Ra).

Physical form

Purified Mouse Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.

Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?