• USA Home
  • MABF434 - Anti-CD45.1 Antibody (mouse), PE-Cy7, clone A20

EMAIL THIS PAGE TO A FRIEND
MABF434 Sigma-Aldrich

Anti-CD45.1 Antibody (mouse), PE-Cy7, clone A20

This mouse monoclonal antibody Anti-CD45.1 (mouse), PE-Cy7, clone A20 validated for use in IP, IHC & Flow Cytometry for the detection of CD45.1.

Synonym: Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45

  •  eCl@ss 32160702

  •  NACRES NA.44

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
packaging   100 µg (MABF434)
application(s)   flow cytometry: suitable
  immunohistochemistry: suitable
  immunoprecipitation (IP): suitable
biological source   mouse
clone   A20, monoclonal
concentration   0.2 mg/mL
isotype   IgG2aκ
shipped in   wet ice
species reactivity   mouse
clone   Monoclonal Antibody
gene symbol   Ly-5
  Ptprc
conjugate   PE-Cy7
Quality Assurance   The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 1 µg of this antibody detected CD45.1 in one million SJL splenocytes.

Description

General description

The A20 antibody reacts with mouse CD45.1, also known as Ly5.1, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The A20 antibody is typically used as a leukocyte marker in Ly5.1 mouse strains: SJL/J, DA, STS/A and RIII. The antibody has been demonstrated to specific for CD45.1 and is not cross-reactive with CD45.2 bearing cells.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?