• USA Home
  • MABN301 - Anti-Peroxiredoxin-5, (PRDX5) Antibody, clone 5 288 2F4

EMAIL THIS PAGE TO A FRIEND
MABN301 Sigma-Aldrich

Anti-Peroxiredoxin-5, (PRDX5) Antibody, clone 5 288 2F4

clone 5 288 2F4, from mouse

Synonym: Peroxiredoxin-5, mitochondrial, Alu corepressor 1, Antioxidant enzyme B166, AOEB166, Liver tissue 2D-page spot 71B, PLP, Peroxiredoxin V, Prx-V, Peroxisomal antioxidant enzyme, TPx type VI, Thioredoxin peroxidase PMP20, Thioredoxin reductase

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

clone   5 288 2F4, monoclonal
biological source   mouse
application(s)   immunohistochemistry: suitable
  western blot: suitable
species reactivity   human
shipped in   wet ice
isotype   IgMκ
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   NP_036226
UniProt accession no.   P30044
Gene Information   human ... PRDX5(25824)

Description

General description

Peroxiredoxin-5 (PRDX5) is a novel mammalian peroxiredoxin, belonging to the family of thiol-dependent peroxidases. It is structurally dissimilar from other peroxiredoxins in that it contains a conserved cysteine 47 residue at the N-terminus and a cysteine 151 residue at the C-terminus which form an intramolecular disulphide bridge and confers unique chemical characteristics to this enzyme. Peroxiredoxins protect cells from apoptosis induced by overexposure to endogenous or exogenous peroxides, such as peroxynitrites, hydrogen peroxide, and alkyl hydroperoxides. Peroxiredoxin 5 is widely expressed in various subcellular regions, including mitochondria, peroxisomes, nucleus, and cytosol.

Immunogen

Recombinant protein corresponding to human Peroxiredoxin-5.

Application

Anti-Peroxiredoxin-5, (PRDX5) Antibody, clone 5 288 2F4 is an antibody against Peroxiredoxin-5, (PRDX5), clone 5 288 2F4 for use in Western Blotting & IHC.

Research Category
Neuroscience

Research Sub Category
Neurodegenerative Diseases

Western Blot Analysis: A 1:1,000 dilution from a representative lot detected Peroxiredoxin-5 (PRDX5) in human brain tissue lysate.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Peroxiredoxin-5 (PRDX5) in human adrenal gland tissue.

Target description

~17 kDa observed

Physical form

Format: Purified

Purified by Ion-Exchange Chromatography

Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Quality

Evaluated by Western Blot in SH-SY5Y cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Peroxiredoxin-5 (PRDX5) in 10 µg of SH-SY5Y cell lysate.

Analysis Note

Control
SH-SY5Y cell lysate

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?