• USA Home
  • MABN521 - Anti-SH3GL1 Antibody, clone K51/1

EMAIL THIS PAGE TO A FRIEND
MABN521 Sigma-Aldrich

Anti-SH3GL1 Antibody, clone K51/1

Detect Endophilin using this mouse monoclonal antibody, Anti-SH3GL1 Antibody & clone K51/1 validated for use in western blotting.

Synonym: Endophilin-A2, Endophilin-2, SH3 domain protein 2B, SH3 domain-containing GRB2-like protein 1, SH3p8

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, SG-SK
clone   K51/1, monoclonal
biological source   mouse
application(s)   western blot: suitable
species reactivity   rat
shipped in   wet ice
isotype   IgG1κ
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   NP_112518
UniProt accession no.   O35964

Description

General description

SH3 domain-containing GRB2-like protein 1(SH3GL1), also known as Endophilin-A2 or Endophilin-2, SH3 domain protein 2B, and SH3p8, is ubiquitously expressed and has been implicated in processes related to endocytosis, and may recruit other proteins to membranes with high curvature. Subcellularly, SH3GL1 is found in the cytoplasm and early endosome membranes. It is also associated with postsynaptic endosomes in hippocampal neurons.

Immunogen

Linear peptide corresponding to rat SH3GL1.

Target description

~ 42 kDa observed

Physical form

Format: Purified

Quality

Evaluated by Western Blotting in L6 cell lysate.

Western Blotting Analysis: 0.45 µg/mL of this antibody detected SH3GL1 in 10 µg of L6 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?