• USA Home
  • MABN727 - Anti-PCDH10 Antibody, clone 4D4.2

EMAIL THIS PAGE TO A FRIEND
MABN727 Sigma-Aldrich

Anti-PCDH10 Antibody, clone 4D4.2

This Anti-PCDH10 Antibody, clone 4D4.2 is validated for use in Western Blotting for the detection of PCDH10.

Synonym: Protocadherin-10

  •  eCl@ss 32160702

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
packaging   100 µg (MABN727)
gene symbol   PCDH10(57575)
format   Purified
molecular weight   ~113 kDa observed
application(s)   Western Blotting
isotype   IgG1κ
clone   Monoclonal Antibody
  4D4.2
concentration   Please refer to lot specific datasheet.
host   Mouse
species reactivity   Human
NCBI accession no.   NP_116586
UniProt accession no.   Q9P2E7
gene symbol   KIAA1400
shipped in   1
Quality Assurance   Evaluated by Western Blotting in human cortex tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected PCDH10 in 10 µg of human cortex tissue lysate.

Description

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

Research Category
Neuroscience

Research Sub Category
Developmental Signaling

General description

Protocadherin-10 is a Single-pass type I membrane protein that is likely involved in calcium-dependent cell-adhesion. Protocadherin-10 is moderately expressed in brain testis and ovaries, with low expression in most other tissues.

Immunogen

GST-tagged recombinant protein corresponding to human PCDH10.

Physical form

Protein G Purified

Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Making Headway
Protocols & Articles

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?