• USA Home
  • MFXSNAP03 - MilliSnap system and Milliflex Plus pump triple head

EMAIL THIS PAGE TO A FRIEND
MFXSNAP03 Millipore

MilliSnap system and Milliflex Plus pump triple head

  •  NACRES NB.79

Purchase

Properties

Related Categories Labware, Liquid Transfer Pumps, Manufacturer Browser, Millipore - Manufacturer Browser, Millipore Filters,
Quality Level   400
mfr. no.   MILLISNAP®
application(s)   bioburden testing: suitable
input   water
Featured Industry   Pharmaceutical
shipped in   ambient

Description

General description

Designed to simplify your workflow, the MilliSnap plug and play system is a hands-free method for separating your Milliflex funnel and attaching the membrane to the Millipore media cassettes. The unique convex shape of the Milliflex membrane combined with the MilliSnap system ensures proper contact between membrane and media.

Legal Information

MILLISNAP is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?