• USA Home
  • MIACN8S01 - Microfil V stainless steel filter support for manifold

EMAIL THIS PAGE TO A FRIEND
MIACN8S01 Millipore

Microfil V stainless steel filter support for manifold

  •  NACRES NB.11

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Millipore - Manufacturer Browser, Millipore Filters, Millipore Filtration Vacuum,
Quality Level   100
mfr. no.   Microfil®
application(s)   bioburden testing: suitable
  water monitoring: suitable
input   sample type beverage(s)
sample type wine
sample type bottled water
sample type food(s)
sample type beer
sample type soft drink
sample type cosmetics
sample type sport drink
sample type cider
Featured Industry   Clinical
Cosmetics
Environmental
Food and Beverages
Industrial QC
shipped in   ambient

Description

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?