• USA Home
  • MSP010022 - Milliflex® Funnel

EMAIL THIS PAGE TO A FRIEND
MSP010022 Millipore

Milliflex® Funnel

Without membrane for bioburden and water analysis with the Milliflex® system

  •  NACRES NB.24

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Millipore - Manufacturer Browser, Millipore Filters, Millipore Filtration Vacuum,
material   clear device
  polypropylene device
  styrene-acrylonitrile (SAN) device
sterility   sterile; ethylene oxide treated
mfr. no.   Milliflex®
application(s)   bioburden testing: suitable
  water monitoring: suitable
input   sample type food(s)
sample type beverage(s)
sample(s) (environmental)
Featured Industry   Clinical
Cosmetics
Environmental
Environmental
Food and Beverages
Industrial QC
shipped in   ambient

Description

General description

Milliflex® funnel without a membrane is a ready-to-use, sterilized filter device for bioburden testing. It is used to perform the culture media growth promotion test with the Milliflex® system.

Application

Milliflex® Funnel without membrane is used in Bioburden testing, water monitoring, and general microbiological analysis of beer, bottled water. They are widely used in the pharmaceutical, cosmetic, and brewing industry.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?