• USA Home
  • MSTHEAD01 - Sterisure head for Milliflex Plus pump

EMAIL THIS PAGE TO A FRIEND
MSTHEAD01 Millipore

Sterisure head for Milliflex Plus pump

Clinical Lab, Food and Beverage, Industrial QC, Environmental Analysis, Laboratory Filtration

  •  NACRES NB.24

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Millipore - Manufacturer Browser, Millipore Filters, Millipore Filtration Vacuum,
material   316L stainless steel
mfr. no.   Milliflex®
application(s)   bioburden testing: suitable
  water monitoring: suitable
input   sample type beer
sample type pharmaceutical(s)
sample type bottled water
sample type cosmetics
Featured Industry   Pharmaceutical
shipped in   ambient

Description

General description

The compact Milliflex PLUS vacuum pump streamlines filtration steps during testing to accelerate QC sampling.

Application

Clinical Lab, Food and Beverage, Industrial QC, Environmental Analysis, Laboratory Filtration

Preparation Note

Sterilization Method
Autoclavable

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?